SRP5257
SMYD2, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
HSKM-B, KMT3C, ZMYND14
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~70 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SMYD2(56950)
General description
SMYD2 (also known as SET and MYND domain-containing protein 2) is a member of the SMYD family of protein methyltransferases. SMYD2 is also known as lysine methyltransferase protein 3C (KMT3C). SMYD2 functions to repress transcription by interacting with the Sin3A histone deacetylase complex and performs methylation of Lys36 in histone H3. The exogenous expression of SMYD2 in mouse fibroblasts decreases cell growth. SMYD2 also function as a putative oncogene by methylating p53 and repressing its tumor suppressive function.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service