SRP5056
PAK6, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
PAK5
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
116-158 nmol/min·mg
mol wt
~110 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PAK6(56924)
General description
PAK6 is a Ser/Thr protein kinase that is a member of the PAK (p21-activated kinase) family that are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. PAK6 is highly expressed in the testis and prostate tissues and interacts with the androgen receptor (AR). In response to androgen, PAK6 cotranslocate into the nucleus with AR. PAK6 is weakly expressed in normal prostate epithelium but the expression is increased in primary and metastatic prostate cancer cells and is further increased in tumors that relapse after androgen deprivation therapy.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ramneet Kaur et al.
The Prostate, 68(14), 1510-1516 (2008-07-22)
PAK6 is a member of the p21-activated kinase (PAK) family of serine/threonine kinases that was originally cloned from prostate cancer (PCa) cells as an androgen receptor interacting protein, but its cellular distribution and functions have not been established. An affinity
F Yang et al.
The Journal of biological chemistry, 276(18), 15345-15353 (2001-03-30)
The androgen receptor (AR) is a hormone-dependent transcription factor that plays important roles in male sexual differentiation and development. Transcription activation by steroid hormone receptors, such as the androgen receptor, is mediated through interaction with cofactors. We recently identified a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service