SRP5018
DAPK2, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
DRP-1, MGC119312
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
115-156 nmol/min·mg
mol wt
~67 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... DAPK2(23604)
General description
DAPK2 or death-associated protein kinase 2 belongs to a family of proapoptotic Ca(2+)/calmodulin-regulated serine/threonine kinases. Overexpression of DAPK2 induces cell apoptosis. DAPK2 has been shown to be a novel Sp1-dependent target gene for E2F1 and Krüppel-like factor 6 (KLF6) in cell death response. Both E2F1 and KLF6 strongly activate the DAPK2 promoter. DAPK2 plays a role in granulopoiesis where it is highly expressed. β-catenin can block anoikis of malignant kidney and intestinal epithelial cells and promote their anchorage-independent growth by down-regulating DAPK2. β-catenin-induced down-regulation of DAPK2 requires the presence of the transcription factor TCF-4.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Britschgi et al.
Oncogene, 27(43), 5706-5716 (2008-06-04)
Death-associated protein kinase 2 (DAPK2) belongs to a family of proapoptotic Ca(2+)/calmodulin-regulated serine/threonine kinases. We recently identified DAPK2 as an enhancing factor during granulocytic differentiation. To identify transcriptional DAPK2 regulators, we cloned 2.7 kb of the 5'-flanking region of the
Hongbing Li et al.
The Journal of biological chemistry, 284(4), 2012-2022 (2008-10-30)
Resistance of solid tumor cells to anoikis, apoptosis induced by cell detachment from the extracellular matrix, is thought to be critical for the ability of these cells to grow anchorage independently within three-dimensional tumor masses and from metastases. Beta-catenin, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service