SRP3113
MIA-2 human
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
Melanoma Inhibitory Activity-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
11.5 kDa
packaging
pkg of 20 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MIA2(117153)
General description
MIA2 is a secreted cytokine and a member of the MIA/OTOR family. Members of this family which also includes MIA, OTOR, and TANGO share a Src homology-3 (SH3)-like domain. MIA2 is predominantly expressed in hepatocytes. Elevated levels of MIA2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity. Recombinant human MIA-2 is a 101 amino acid, 11.5 kDa protein comprising the MIA homologous N-terminal region of the full length MIA-2 protein.
Biochem/physiol Actions
MIA2 is a secreted cytokine and a member of the MIA/OTOR family. Recombinant human MIA-2 is a 101 amino acid, 11.5 kDa protein comprising the MIA homologous N-terminal region of the full length MIA-2 protein.
Sequence
MLESTKLLAD LKKCGDLECE ALINRVSAMR DYRGPDCRYL NFTKGEEISV YVKLAGERED LWAGSKGKEF GYFPRDAVQI EEVFISEEIQ MSTKESDFLC L
Physical form
Lyophilized from 10 mM Sodium Phosphate, pH 7.5 + 0.5 mM DTT.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A K Bosserhoff et al.
Histology and histopathology, 17(1), 289-300 (2002-01-30)
Despite its ambiguous name the protein melanoma-inhibitory-activity (MIA) was identified as a key molecule involved in progression and metastasis of malignant melanomas. Therefore, in this review we intend to update the current knowledge on expression patterns, transcriptional regulation, function and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service