Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4059

Sigma-Aldrich

Elagolix sodium

≥98% (HPLC)

Synonym(s):

(R)-4-{2-[5-(2-Fluoro-3-methoxyphenyl)-3-(2-fluoro-6-[trifluoromethyl]benzyl)-4-methyl-2,6-dioxo-3,6-dihydro-2H-pyrimidin-1-yl]-1-phenylethylamino}butyric acid sodium salt, Elagolix sodium salt, NBI 56418, NBI-56418, NBI56418

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H29F5N3O5·Na
CAS Number:
Molecular Weight:
653.57
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=C(C2=C(C(OC)=CC=C2)F)C(N(C(N1CC3=C(C=CC=C3F)C(F)(F)F)=O)C[C@@H](C4=CC=CC=C4)NCCCC(O[Na])=O)=O

Biochem/physiol Actions

Orally active human gonadotropin releasing hormone (GnRH)/luteinizing hormone releasing hormone (LHRH) receptor (GnRHR, LHRHR) antagonist.
Elagolix is an orally active, potent, high-affinity human gonadotropin-releasing hormone (GnRH)/luteinizing hormone-releasing hormone (LH-RH) receptor (GnRHR, LHRHR) antagonist (Ki = 0.9 nM/human, 3.3 nM/monkey, 4.4 µM/rat) that effectively blocks GnRH-induced inositol phosphate production in RBL-1 hGnRH-R transfectants (IC50 = 1.5 nM against 6 nM GnRH). Oral administration in castrated male cynomolgus macaques effectively reduces circulating LH in a reversible manner in vivo (75% reduction 8h post 30 mg/kg p.o.).

Caution

Hygroscopic

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service