biological source
synthetic
form
liquid
species reactivity
human, mouse, rat
concentration
200 μg/mL
shipped in
wet ice
storage temp.
−20°C
Immunogen
20 amino acids near the amino terminus of human JMJD1C.
Application
Used as a blocking peptide in immunoblotting applications.
Linkage
This peptide blocks the action of antibody SAB3500094.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Related product
Product No.
Description
Pricing
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service