Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SBP3377

Sigma-Aldrich

ECSIT Blocking Peptide for PRS3377

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
SGD 591.00

SGD 591.00


Estimated to ship onApril 24, 2025Details



Select a Size

Change View
100 μL
SGD 591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

SGD 591.00


Estimated to ship onApril 24, 2025Details


biological source

synthetic

form

liquid

species reactivity

mouse, human

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1410765HPA028897SAB4501813
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

bovine, mouse, rabbit, horse, human, rat, guinea pig, dog

species reactivity

human

species reactivity

human

species reactivity

rat, mouse, human

Gene Information

human ... SMAD5(4090)

Gene Information

human ... SMAD9(4093)

Gene Information

human ... SMAD7(4092)

Gene Information

human ... SMAD3(4088)

Immunogen

14 amino acids near the carboxy terminus of human ECSIT.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody PRS3377.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service