SAB6100007
Anti-IFNg Reference Antibody

Synonym(s):
IFG
About This Item
recombinant
expressed in CHO cells
Quality Level
conjugate
unconjugated
antibody product type
primary antibodies
clone
recombinant monoclonal
form
lyophilized
species reactivity
human, macaque (Cynomolgus), mouse
concentration
1 mg/mL
technique(s)
ELISA: 0.0001-10 μg/mL
optimum pH
6.2
pH range
5.5-6.11
immunogen sequence
MCHQQLVISWFSLVFLASPLVAIWELKKDVYVVELDWYPDAPGEMVVLTCDTPEEDGITWTLDQSSEVLGSGKTLTIQVKEFGDAGQYTCHKGGEVLSHSLLLLHKKEDGIWSTDILKDQKEPKNKTFLRCEAKNYSGRFTCWWLTTISTDLTFSVKSSRGSSDPQGVTCGAATLSAERVRGDNKEYEYSVECQEDSACPAAEESLPIEVMVDAVHKLKYENYTSSFFIRDIIKPDPPKNLQLKPLKNSRQVEVSWEYPDTWSTPHSYFSLTFCVQVQGKSKREKKDRVFTDKTSATVICRKNASISVRAQDRYYSSSWSEWASVPCS
Protein ID accession no.
UniProt accession no.
storage temp.
-65 to -96°C
target post-translational modification
unmodified
Gene Information
human ... IFNG(3458)
General description
Immunogen
Packaging
Reconstitution
Storage and Stability
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service