Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702398

Sigma-Aldrich

Anti-CENPC Antibody, clone 5E3K5, Rabbit Monoclonal

Synonym(s):

CENP-C, CENPC1, MIF2, hcp-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5E3K5, monoclonal

form

liquid

mol wt

107 kDa

species reactivity

human, rat, rat

concentration

1.85 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

VLDTSYALIGETVNNYRSTKYEMYSKNAEKPSRSKRTIKQKQRRKFMAKPAEEQLDVGQSKDENIHTSHITQDEFQRNSDRNMEEHEEMGNDCVSKKQMPPVGSKKSSTRKDKEESKKKRFSSESKNKLVPEEVTSTVTKS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CENPC(1060)

General description

Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12. [provided by RefSeq, Jul 2008]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 380-520 of human CENPC (Q03188).

Application

WB, IF/ICC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service