Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5702239

Sigma-Aldrich

[KO Validated]Anti-CDX2 Antibody, clone 4Y1O3, Rabbit Monoclonal

Synonym(s):

CDX2,CDX-3,CDX2/AS,CDX3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

4Y1O3, monoclonal

form

liquid

mol wt

38 kDa

species reactivity

mouse, mouse

concentration

0.50 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDX2(1045)

General description

This gene is a member of the caudal-related homeobox transcription factor gene family. The encoded protein is a major regulator of intestine-specific genes involved in cell growth an differentiation. This protein also plays a role in early embryonic development of the intestinal tract. Aberrant expression of this gene is associated with intestinal inflammation and tumorigenesis. [provided by RefSeq, Jan 2012]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant protein of human CDX2.

Application

WB, IHC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service