Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5702190

Sigma-Aldrich

Anti-TAX1BP3 Antibody, clone 9T0I1, Rabbit Monoclonal

Synonym(s):

TIP-1, TIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

9T0I1, monoclonal

form

liquid

mol wt

14 kDa

species reactivity

rat, human, rat

concentration

3.00 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSYIPGQPVTAVVQRVEIHKLRQGENLILGFSIGGGIDQDPSQNPFSEDKTDKGIYVTRVSEGGPAEIAGLQIGDKIMQVNGWDMTMVTHDQARKRLTKR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAX1BP3(30851)

General description

This gene encodes a small, highly conserved protein with a single PDZ domain. PDZ (PSD-95/Discs large/ZO-1 homologous) domains promote protein-protein interactions that affect cell signaling, adhesion, protein scaffolding, and receptor and ion transporter functions. The encoded protein interacts with a large number of target proteins that play roles in signaling pathways; for example, it interacts with Rho A and glutaminase L and also acts as a negative regulator of the Wnt/beta-catenin signaling pathway. This protein was first identified as binding to the T-cell leukaemia virus (HTLV1) Tax oncoprotein. Overexpression of this gene has been implicated in altered cancer cell adhesion, migration and metastasis. The encoded protein also modulates the localization and density of inwardly rectifying potassium channel 2.3 (Kir2.3). To date, this protein has been shown to play a role in cell proliferation, development, stress response, and polarization. Alternative splicing results in multiple transcript variants encoding distinct isoforms.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TAX1BP3 (O14907).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service