Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB5702051

Sigma-Aldrich

Anti-DEK Antibody, clone 2C2V8, Rabbit Monoclonal

Synonym(s):

D6S231E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

2C2V8, monoclonal

form

liquid

mol wt

50 kDa

species reactivity

human, rat

concentration

0.50 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

SLIVEGKREKKKVERLTMQVSSLQREPFTIAQGKGQKLCEIERIHFFLSKKKTDELRNLHKLLYNRPGTVSSLKKNVGQFSGFPFEKGSVQYKKKEEMLKKFRNAMLKSICEVLDLERSGVNSELVKRILNFLMHPKPSGKPLPKSKKTC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DEK(7913)

General description

This gene encodes a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2008]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 51-200 of human DEK (P35659).

Application

WB, IHC, IF/ICC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service