Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5700546

Sigma-Aldrich

Anti-Phospho-PPP2CA/PPP2CB-Y307 antibody produced in rabbit

Synonym(s):

PPP2CA, PPP2CB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

1.91mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr307)

Gene Information

human ... PPP2CA(5515)

Immunogen

A phospho specific peptide corresponding to residues surrounding Y307 of human PPP2CA/PPP2CB

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service