SAB5500154
Anti-Nucleophosmin antibody, Rabbit monoclonal
recombinant, expressed in proprietary host, clone SP236, affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
rabbit
Quality Level
recombinant
expressed in proprietary host
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
SP236, monoclonal
species reactivity
human (tested)
species reactivity (predicted by homology)
bovine, rat, chicken, mouse
technique(s)
flow cytometry: 1:400
immunoblotting: 1:400
immunohistochemistry: 1:100
isotype
IgG
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... NPM1(4869)
Related Categories
General description
Nucleophosmin (NPM) is a nucleolar protein for regulating ribosome biosynthesis and centrosome duplication. The NPM gene is frequently rearranged in various types of lymphomas such as anaplastic lymphoma kinase (ALK). A C-terminal mutation occurs in acute myelogenous leukemia (AML) and prevents localization to the nucleolus, resulting in cytoplasmic distribution. NPM is frequently overexpressed in diverse solid tumors such as carcinoma of the lung and breast, colon, prostate, stomach, lymphoid and myeloid tumors.
The NPM1 (nucleophosmin 1) gene is mapped to human chromosome 5q35.1. Nucleophosmin is a predominant phospho-nucleolar protein, that is mostly, localized to the nucleus or cytoplasm.
Immunogen
Synthetic peptide derived from human nucleophosmin.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Expression of the cytoplasmic NPM1 mutant (NPMc+) causes the expansion of hematopoietic cells in zebrafish
Bolli N, et al.
Blood (2010)
Expression of the cytoplasmic NPM1 mutant (NPMc+) causes the expansion of hematopoietic cells in zebrafish
Bolli N, et al.
Blood (2010)
Kai Yang et al.
Nature communications, 7, 13599-13599 (2016-11-26)
The nucleolus has been recently described as a stress sensor. The nucleoplasmic translocation of nucleolar protein nucleophosmin (NPM1) is a hallmark of nucleolar stress; however, the causes of this translocation and its connection to p53 activation are unclear. Using single
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service