Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200006

Sigma-Aldrich

Monoclonal Anti-CRYAA antibody produced in mouse

clone 1H3.B8, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-Acry1, Anti-Alpha A Crystallin, Anti-CRYA1, Anti-CRYAA, Anti-Heat Shock protein beta4, Anti-HspB4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3.B8, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 20 kDa

species reactivity

bovine, rat, mouse, human

concentration

1 mg/mL

technique(s)

indirect ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRYAA(1409)

Specificity

Detects α-crystallin at ~20 kDa. Does not cross-react with αB-crystallin, βL-crystallin, βH- crystallin, γ-crystallin, Hsp25, Hsp27 or Hsp47 proteins.

Immunogen

Native Alpha Crystallin

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.2, in 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiwani Sharma et al.
Molecular vision, 24, 801-817 (2019-02-05)
Pseudoexfoliation (PEX) syndrome is an age-related progressive disease of the extracellular matrix with ocular manifestations. PEX is clinically diagnosed by the presence of extracellular exfoliative deposits on the anterior surface of the ocular lens. PEX syndrome is a major risk
Xiang-Jia Zhu et al.
Journal of cellular and molecular medicine, 22(2), 1118-1126 (2017-10-11)
Post-translational modifications in lens proteins are key causal factors in cataract. As the most abundant post-translational modification in the lens, racemization may be closely related to the pathogenesis of cataract. Racemization of αA-crystallin, a crucial structural and heat shock protein
Andrea Schlotterer et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(3), 4141-4153 (2018-11-30)
The aim of this study was to evaluate whether damage to the neurovascular unit in diabetes depends on reactive metabolites such as methylglyoxal (MG), and to assess its impact on retinal gene expression. Male Wistar rats were supplied with MG
Jason Tchieu et al.
Cell stem cell, 21(3), 399-410 (2017-09-09)
Directing the fate of human pluripotent stem cells (hPSCs) into different lineages requires variable starting conditions and components with undefined activities, introducing inconsistencies that confound reproducibility and assessment of specific perturbations. Here we introduce a simple, modular protocol for deriving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service