SAB4700940
Monoclonal Anti-CD64Biotin antibody produced in mouse
clone 10.1
Synonym(s):
FcRI, FcgR1A, IGFR1
About This Item
biological source
mouse
antibody product type
primary antibodies
clone
10.1, monoclonal
form
buffered aqueous solution
species reactivity
human, nonhuman primates
concentration
1 mg/mL
technique(s)
flow cytometry: 6 μg/mL
accession no.
NP_000557.1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... FCGR1A(2209)
Related Categories
General description
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service