Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502673

Sigma-Aldrich

Anti-MRPL12 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

39S ribosomal protein L12, 5c5-2, MRP-L12, mitochondrial L12mt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL12(6182)

General description

Anti-MRPL12 Antibody detects endogenous levels of total MRPL12 protein.
MRPL12 (mitochondrial ribosomal protein L12) encodes a 21kDa subunit of mitochondrial ribosome. The protein is mapped on chromosome 17 (q25-qter). It is highly expressed in the colon.

Immunogen

The antiserum was produced against synthesized peptide derived from human MRPL12.

Immunogen Range: 31-80

Application

Anti-MRPL12 antibody produced in rabbit is suitable for capture ELISA and westen blot.

Biochem/physiol Actions

MRPL12 (mitochondrial ribosomal protein) is involved in the mitochondrial translation. It directly interacts with the mitochondrial RNA polymerase (POLRMT) and triggers the mitochondrial transcriptional process. Research shows that the interaction between MRPL12 and POLRMT indicates its participation in the regulatory mechanism that coordinates mitochondrial transcription with translation and/or ribosome biogenesis during human mitochondrial gene expression. Study shows that MRPL12 may be involved in the differentiation of colonic epithelial cells and its reduced mitochondrial activity have been shown to be linked with tumor formation. It has been reported that mutated expression of MRPL12 associates with the growth retardation, neurological deterioration and mitochondrial translation deficiency.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valérie Serre et al.
Biochimica et biophysica acta, 1832(8), 1304-1312 (2013-04-23)
Multiple respiratory chain deficiencies represent a common cause of mitochondrial diseases and are associated with a wide range of clinical symptoms. We report a subject, born to consanguineous parents, with growth retardation and neurological deterioration. Multiple respiratory chain deficiency was
L Marty et al.
Genomics, 41(3), 453-457 (1997-05-01)
Mitochondrial activity requires the expression of nuclear genes, whose products are part of multiproteic complexes leading to ATP production and delivery. We recently characterized a growth-activated mRNA encoding the human mitochondrial ribosomal MRPL12 protein, which is thought to act as
Zhibo Wang et al.
The Journal of biological chemistry, 282(17), 12610-12618 (2007-03-06)
The core human mitochondrial transcription machinery comprises a single subunit bacteriophage-related RNA polymerase, POLRMT, the high mobility group box DNA-binding protein h-mtTFA/TFAM, and two transcriptional co-activator proteins, h-mtTFB1 and h-mtTFB2 that also have rRNA methyltransferase activity. Recapitulation of specific initiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service