Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501805

Sigma-Aldrich

Anti-SMAD2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

JV18-1, MADH2, MADR2, Mad-related protein 2, Mothers against DPP homolog 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD2(4087)

General description

Anti-SMAD2 Antibody detects endogenous levels of total SMAD2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Smad2.

Immunogen Range: 221-270

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Yang et al.
Oncology research, 21(6), 345-352 (2013-01-01)
TGF-β/Smad signaling induces epithelial-mesenchymal transition (EMT) and tumor metastasis. As essential mediators in this pathway, Smad2 and Smad3 have been extensively studied and found to promote EMT and the subsequent mobility as well as invasiveness of lung cancer cells. In
Hye Sook Min et al.
Laboratory investigation; a journal of technical methods and pathology, 94(6), 598-607 (2014-04-02)
Dipeptidyl peptidase IV (DPPIV) is an exopeptidase that modulates the function of several substrates, among which insulin-releasing incretin hormones are the most well known. DPPIV also modulate substrates involved in inflammation, cell migration, and cell differentiation. Although DPPIV is highly
Minghua Wu et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(4), 1010-1021 (2014-04-24)
Systemic sclerosis (SSc) is a chronic autoimmune disease clinically manifesting as progressive fibrosis of the skin and internal organs. Recent microarray studies demonstrated that cadherin 11 (Cad-11) expression is increased in the affected skin of patients with SSc. The purpose
Julien Dubrulle et al.
eLife, 4 (2015-04-15)
Morphogen gradients expose cells to different signal concentrations and induce target genes with different ranges of expression. To determine how the Nodal morphogen gradient induces distinct gene expression patterns during zebrafish embryogenesis, we measured the activation dynamics of the signal
Christine Bruun et al.
Diabetologia, 57(12), 2546-2554 (2014-09-28)
Impairment of beta cell mass and function is evident in both type 1 and type 2 diabetes. In healthy physiological conditions pancreatic beta cells adapt to the body's increasing insulin requirements by proliferation and improved function. We hypothesised that during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service