Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501181

Sigma-Aldrich

Anti-DNAI1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CILD1, ICS, MGC26204, PCD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 79 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAI1(27019)

General description

Anti-DNAI1 Antibody detects endogenous levels of total DNAI1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human DNAI1.

Immunogen Range: 211-260

Application

Anti-DNAI1 antibody produced in rabbit is suitable for indirect ELISA and western blot.

Biochem/physiol Actions

DNAI1 (Dynein axonemal intermediate chain 1) encodes an intermediate of outer dynein arms (ODA) proteins of cilia. Mutation in DNAI1 gene causes a trilogy syndrome named as Kartagener syndrome (KS), a rare recessive hereditary disorder named primary ciliary dyskinesia (PCD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenglan Dong et al.
The Journal of cell biology, 204(2), 203-213 (2014-01-15)
Axonemal dynein complexes are preassembled in the cytoplasm before their transport to cilia, but the mechanism of this process remains unclear. We now show that mice lacking Pih1d3, a PIH1 domain-containing protein, develop normally but manifest male sterility. Pih1d3(-/-) sperm
Maimoona A Zariwala et al.
American journal of respiratory and critical care medicine, 174(8), 858-866 (2006-07-22)
Primary ciliary dyskinesia (PCD) is a rare, usually autosomal recessive, genetic disorder characterized by ciliary dysfunction, sino-pulmonary disease, and situs inversus. Disease-causing mutations have been reported in DNAI1 and DNAH5 encoding outer dynein arm (ODA) proteins of cilia. We analyzed
Mike Failly et al.
Respiration; international review of thoracic diseases, 76(2), 198-204 (2008-04-25)
Primary ciliary dyskinesia (PCD) is a rare recessive hereditary disorder characterized by dysmotility to immotility of ciliated and flagellated structures. Its main symptoms are respiratory, caused by defective ciliary beating in the epithelium of the upper airways (nose, bronchi and
C Guichard et al.
American journal of human genetics, 68(4), 1030-1035 (2001-03-07)
Kartagener syndrome (KS) is a trilogy of symptoms (nasal polyps, bronchiectasis, and situs inversus totalis) that is associated with ultrastructural anomalies of cilia of epithelial cells covering the upper and lower respiratory tracts and spermatozoa flagellae. The axonemal dynein intermediate-chain
Kyeong Jee Cho et al.
PLoS genetics, 14(3), e1007316-e1007316 (2018-03-31)
Zinc finger MYND-type-containing 10 (ZMYND10), a cytoplasmic protein expressed in ciliated cells, causes primary ciliary dyskinesia (PCD) when mutated; however, its function is poorly understood. Therefore, in this study, we examined the roles of ZMYND10 using Zmynd10-/-mice exhibiting typical PCD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service