Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4300560

Sigma-Aldrich

Anti-PTPN11 (Ab-580) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BPTP3 antibody produced in rabbit, Anti-CFC antibody produced in rabbit, Anti-MGC14433 antibody produced in rabbit, Anti-NS1 antibody produced in rabbit, Anti-protein tyrosine phosphatase, non-receptor type 11 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~72 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-V-Y-E-N)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPN11(5781)

Immunogen

Peptide sequence around aa. 578-582 (R-V-Y-E-N), according to the protein PTPN11.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Acts downstream of various receptor and cytoplasmic protein tyrosine kinases to participate in the signal transduction from the cell surface to the nucleus.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toru Mitsumori et al.
Experimental hematology, 42(9), 783-792 (2014-05-28)
The hypoxic microenvironment of the bone marrow, known as the hypoxic niche, supports hematopoietic stem cell quiescence and maintains long-term repopulation activity. Hypoxia also affects the expansion of progenitor cells and enhances erythropoiesis and megakaryopoiesis. In contrast to the known
Diego Allonso et al.
PloS one, 9(11), e113634-e113634 (2014-11-21)
Dengue virus (DENV) represents a major threat to public health worldwide. Early DENV diagnosis should not only detect the infection but also identify patients with a higher likelihood to develop severe cases. Previous studies have suggested the potential for NS1
Song Wang et al.
Journal of virology, 88(15), 8375-8385 (2014-05-16)
Although alteration in host cellular translation machinery occurs in virus-infected cells, the role of such alteration and the precise pathogenic processes are not well understood. Influenza A virus (IAV) infection shuts off host cell gene expression at transcriptional and translational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service