Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300097

Sigma-Aldrich

Anti-phospho-CDC25C (pSer216) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CDC25 antibody produced in rabbit, Anti-cell division cycle 25 homolog C (S. pombe) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~60 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(S-P-SP-M-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer216)

Gene Information

human ... CDC25C(995)

Immunogen

Peptide sequence around phosphorylation site of serine 216 (S-P-S(p)-M-P), according to the protein CDC25C.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

cdc25C is highly conserved during evolution and it plays a key role in the regulation of cell division. The encodedprotein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclinB-bound CDC2 and triggers entry into mitosis. It is also thought to suppress p53-induced growth arrest. Multiplealternatively spliced transcript variants of this gene have been described, however, the full-length nature of many ofthem is not known.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service