Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500980

Sigma-Aldrich

Anti-CTRP6 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Antibody was raised against a 15 amino acid synthetic peptide from near the center of human CTRP6.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500980.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsu-Shuen Tsao et al.
European journal of pharmacology, 440(2-3), 213-221 (2002-05-15)
Adipocyte complement-related protein of 30 kDa (ACRP30) is a secreted serum protein expressed exclusively in differentiated adipocytes. Recent studies have indicated that its expression and serum levels are reduced in humans and animals with obesity and insulin resistance. Metabolic studies
Giamila Fantuzzi
The Journal of allergy and clinical immunology, 115(5), 911-919 (2005-05-04)
White adipose tissue is no longer considered an inert tissue mainly devoted to energy storage but is emerging as an active participant in regulating physiologic and pathologic processes, including immunity and inflammation. Macrophages are components of adipose tissue and actively
L Shapiro et al.
Current biology : CB, 8(6), 335-338 (1998-03-25)
ACRP30--adipocyte complement-related protein of 30 kDa or AdipoQ--is an abundant serum protein, secreted exclusively from fat cells, which is implicated in energy homeostasis and obesity [1,2]. ACRP30 is a close homologue of the complement protein C1q, which is involved in
Guang W Wong et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(28), 10302-10307 (2004-07-03)
Biochemical, genetic, and animal studies in recent years have established a critical role for the adipokine Acrp30/adiponectin in controlling whole-body metabolism, particularly by enhancing insulin sensitivity in muscle and liver, and by increasing fatty acid oxidation in muscle. We describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service