Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501314

Sigma-Aldrich

Anti-SUMF1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AAPA3037, Anti-FGE, Anti-MGC131853, Anti-MGC150436, sulfatase modifying factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUMF1(285362)

General description

Sulfatase modifying factor 1 (SUMF1) or formylglycine-generating enzyme (FGE) is a 41kDa glycoprotein. It is localized at the lumen of the endoplasmic reticulum. The protein consists of an amino-terminal domain and a core domain with the active site. The gene encoding SUMF1 is localized on human chromosome 3p26.1 and contains nine exons.

Immunogen

Peptide with sequence CETLNPKGPPSGKDR, from the internal region of the protein sequence according to NP_877437.2.

Biochem/physiol Actions

Sulfatase modifying factor 1 (SUMF1) converts a highly conserved cysteine in the catalytic domain of sulfatase to form C α-formylglycine (FGly) by post-translational modification. Mutations in the gene encoding this protein have been associated with multiple sulfatase deficiency.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple sulfatase deficiency with neonatal manifestation.
Garavelli L
Italian Journal of Pediatrics (2014)
Eukaryotic formylglycine-generating enzyme catalyses a monooxygenase type of reaction.
Peng J
FEBS Journal, 282(17) (2015)
Natural disease history and characterisation of SUMF1 molecular defects in ten unrelated patients with multiple sulfatase deficiency.
Sabourdy F
Orphanet Journal of Rare Diseases (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service