Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2105482

Sigma-Aldrich

Anti-RGS10, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-2310010N19Rik, Anti-MGC129414

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

human, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS10(6001)
mouse ... Rgs10(67865)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal of human Rgs10

Biochem/physiol Actions

Rgs10 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Rgs10 associates specifically with the activated forms of the G protein subunits G(i)-alpha and G(z)-alpha but fails to interact with the structurally and functionally distinct G(s)-alpha subunit. Activity of Rgs10 on G(z)-alpha is inhibited by palmitoylation of the G-protein.

Sequence

Synthetic peptide located within the following region: MFTRAVSRLSRKRPPSDIHDGDGSSSSGHQSLKSTAKWASSLENLLEDPE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service