Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2105242

Sigma-Aldrich

Anti-NOTO, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Flh, Anti-MmNot, Anti-Not, Anti-tc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

26 kDa

species reactivity

human, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NOTO(344022)
mouse ... Noto(384452)

Immunogen

Synthetic peptide directed towards the n terminal region of mouse Noto

Biochem/physiol Actions

Noto is a transcription regulator acting downstream of both FOXA2 and T during notochord development.Noto is required for node morphogenesis.Noto is essential for cilia formation in the posterior notochord (PNC) and for left-right patterning; acts upstream of FOXJ1 and RFX3 in this process and is required for the expression of various components important for axonemal assembly and function. Plays a role in regulating axial versus paraxial cell fate.

Sequence

Synthetic peptide located within the following region: VQPGSLRPCPGAVSPVVPRRLARGRLESSFSVEAILARPKTRELAATSLP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiraporn Jirakkakul et al.
FEMS microbiology letters, 362(2), 1-8 (2015-02-12)
Iron is an essential element for life. However, the iron overload can be toxic. Here, we investigated the significant increase of tenellin and iron-tenellin complex production in ferricrocin-deficient mutants of Beauveria bassiana. Our chemical analysis indicated that the ferricrocin-deficient mutants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service