Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

SAB2101216

Sigma-Aldrich

Anti-KCNJ12 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ14167, Anti-IRK2, Anti-KCNJN1, Anti-Kir2.2, Anti-Potassium inwardly-rectifying channel, subfamily J, member 12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

49 kDa

species reactivity

mouse, rat, human, guinea pig, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KCNJ12(3768)

Immunogen

Synthetic peptide directed towards the middle region of human KCNJ12

Biochem/physiol Actions

KCNJ12 is an inwardly rectifying K+ channel which may be blocked by divalent cations. This protein is thought to be one of multiple inwardly rectifying channels which contribute to the cardiac inward rectifier current (IK1). This gene is located within the Smith-Magenis syndrome region on chromosome 17. This gene encodes an inwardly rectifying K+ channel which may be blocked by divalent cations. This protein is thought to be one of multiple inwardly rectifying channels which contribute to the cardiac inward rectifier current (IK1). The gene is located within the Smith-Magenis syndrome region on chromosome 17. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-486 BC027982.1 12-497 487-488 AC068418.5 101651-101652 489-2455 BC027982.1 500-2466 2456-2483 DA115102.1 132-159 2484-2485 AC068418.5 141899-141900 2486-2889 DA115102.1 161-564 2890-3454 BM799671.1 118-682 3455-3457 AC068418.5 142870-142872 3458-4692 AK024229.1 926-2160 4693-5230 AK024229.1 2163-2700

Sequence

Synthetic peptide located within the following region: KDLVENKFLLPSANSFCYENELAFLSRDEEDEADGDQDGRSRDGLSPQAR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service