Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1412297

Sigma-Aldrich

ANTI-HEYL antibody produced in mouse

clone 1E2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HEYL, HRT3, MGC12623, bHLHb33

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E2, monoclonal

form

buffered aqueous solution

mol wt

antigen 33.33 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HEYL(26508)

General description

This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. (provided by RefSeq)

Immunogen

HEYL (NP_055386, 1 a.a. ~ 70 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MKRPKEPSGSDGESDGPIDVGQEGQLSQMARPLSTPSSSQMQARKKRRGIIEKRRRDRINSSLSELRRLV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service