Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1411510

Sigma-Aldrich

Anti-GNPAT antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DAP-AT, DAPAT, DHAPAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 77.2 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNPAT(8443)

General description

This gene encodes an enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in this gene are associated with rhizomelic chondrodysplasia punctata. (provided by RefSeq)

Immunogen

GNPAT (NP_055051.1, 1 a.a. ~ 680 a.a) full-length human protein.

Sequence
MESSSSSNSYFSVGPTSPSAVVLLYSKELKKWDEFEDILEERRHVSDLKFAMKCYTPLVYKGITPCKPIDIKCSVLNSEEIHYVIKQLSKESLQSVDVLREEVSEILDEMSHKLRLGAIRFCAFTLSKVFKQIFSKVCVNEEGIQKLQRAIQEHPVVLLPSHRSYIDFLMLSFLLYNYDLPVPVIAAGMDFLGMKMVGELLRMSGAFFMRRTFGGNKLYWAVFSEYVKTMLRNGYAPVEFFLEGTRSRSAKTLTPKFGLLNIVMEPFFKREVFDTYLVPISISYDKILEETLYVYELLGVPKPKESTTGLLKARKILSENFGSIHVYFGDPVSLRSLAAGRMSRSSYNLVPRYIPQKQSEDMHAFVTEVAYKMELLQIENMVLSPWTLIVAVLLQNRPSMDFDALVEKTLWLKGLTQAFGGFLIWPDNKPAEEVVPASILLHSNIASLVKDQVILKVDSGDSEVVDGLMLQHITLLMCSAYRNQLLNIFVRPSLVAVALQMTPGFRKEDVYSCFRFLRDVFADEFIFLPGNTLKDFEEGCYLLCKSEAIQVTTKDILVTEKGNTVLEFLVGLFKPFVESYQIICKYLLSEEEDHFSEEQYLAAVRKFTSQLLDQGTSQCYDVLSSDVQKNALAACVRLGVVEKKKINNNCIFNVNEPATTKLEEMLGCKTPIGKPATAKL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service