Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1410973

Sigma-Aldrich

Anti-NUP54 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC13407

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NUP54(53371)

General description

The nuclear envelope creates distinct nuclear and cytoplasmic compartments in eukaryotic cells. It consists of two concentric membranes perforated by nuclear pores, large protein complexes that form aqueous channels to regulate the flow of macromolecules between the nucleus and the cytoplasm. These complexes are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. This gene encodes a member of the phe-gly (FG) repeat-containing nucleoporin subset. (provided by RefSeq)

Immunogen

NUP54 (NP_059122.2, 1 a.a. ~ 507 a.a) full-length human protein.

Sequence
MAFNFGAPSGTSGTAAATAAPAGGFGGFGTTSTTAGSAFSFSAPTNTGTTGLFGGTQNKGFGFGTGFGTTTGTSTGLGTGLGTGLGFGGFNTQQQQQTTLGGLFSQPTQAPTQSNQLINTASALSAPTLLGDERDAILAKWNQLQAFWGTGKGYFNNNIPPVEFTQENPFCRFKAVGYSCMPSNKDEDGLVVLVFNKKETEIRSQQQQLVESLHKVLGGNQTLTVNVEGTKTLPDDQTEVVIYVVERSPNGTSRRVPATTLYAHFEQANIKTQLQQLGVTLSMTRTELSPAQIKQLLQNPPAGVDPIIWEQAKVDNPDSEKLIPVPMVGFKELLRRLKVQDQMTKQHQTRLDIISEDISELQKNQTTSVAKIAQYKRKLMDLSHRTLQVLIKQEIQRKSGYAIQADEEQLRVQLDTIQGELNAPTQFKGRLNELMSQIRMQNHFGAVRSEERYYIDADLLREIKQHLKQQQEGLSHLISIIKDDLEDIKLVEHGLNETIHIRGGVFS

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service