Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1409951

Sigma-Aldrich

Anti-WISP2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CCN5, CT58, CTGF-L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WISP2(8839)

General description

This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like (CT) domain. The encoded protein lacks the CT domain which is implicated in dimerization and heparin binding. It is 72% identical to the mouse protein at the amino acid level. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. Its expression in colon tumors is reduced while the other two WISP members are overexpressed in colon tumors. It is expressed at high levels in bone tissue, and may play an important role in modulating bone turnover. (provided by RefSeq)

Immunogen

WISP2 (NP_003872.1, 1 a.a. ~ 250 a.a) full-length human protein.

Sequence
MRGTPKTHLLAFSLLCLLSKVRTQLCPTPCTCPWPPPRCPLGVPLVLDGCGCCRVCARRLGEPCDQLHVCDASQGLVCQPGAGPGGRGALCLLAEDDSSCEVNGRLYREGETFQPHCSIRCRCEDGGFTCVPLCSEDVRLPSWDCPHPRRVEVLGKCCPEWVCGQGGGLGTQPLPAQGPQFSGLVSSLPPGVPCPEWSTAWGPCSTTCGLGMATRVSNQNRFCRLETQRRLCLSRPCPPSRGRSPQNSAF

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service