Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408905

Sigma-Aldrich

Anti-NRG4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp779N0541, DKFZp779N1944, HRG4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 12.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRG4(145957)

Immunogen

NRG4 (NP_612640.1, 1 a.a. ~ 115 a.a) full-length human protein.

Sequence
MPTDHEEPCGPSHKSFCLNGGLCYVIPTIPSPFCRCVENYTGARCEEVFLPGSSIQTKSNLFEAFVALAVLVTLIIGAFYFLCRKGHFQRASSVQYDINLVETSSTSAHHSHEQH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proximal and distal 15q25.2 microdeletions-genotype-phenotype delineation of two neurodevelopmental susceptibility loci.
Doelken SC
American Journal of Medical Genetics. Part A, 161A, 218-218 (2013)
Guo-Xiao Wang et al.
Nature medicine, 20(12), 1436-1443 (2014-11-18)
Brown fat activates uncoupled respiration in response to cold temperature and contributes to systemic metabolic homeostasis. To date, the metabolic action of brown fat has been primarily attributed to its role in fuel oxidation and uncoupling protein 1 (UCP1)-mediated thermogenesis.
Jessica K Bernard et al.
The Journal of biological chemistry, 287(47), 39850-39858 (2012-10-04)
Expression of the ErbB4 tyrosine kinase is elevated in colonic epithelial cells during inflammatory bowel disease, whereas ErbB4 overexpression in cultured colonocytes blocks TNF-induced apoptosis in a ligand-dependent manner. Together, these observations suggest that ErbB4 induction may be a protective
A A Memon et al.
British journal of cancer, 91(12), 2034-2041 (2004-12-08)
The epidermal growth factor system has been associated to prognosis in patients with bladder cancer based mainly on the expression of the epidermal growth factor (EGF) receptor 1 (EGFR) and HER2 and their activating ligands. Since limited information exists concerning
Nandini V L Hayes et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(11), 3147-3155 (2007-06-05)
The neuregulin (NRG) 1, 2, and 3 genes undergo extensive alternative mRNA splicing, which results in variants that show structural and functional diversity. The aims of this study were to establish whether the fourth member of this family, NRG4, is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service