Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1408102

Sigma-Aldrich

Anti-ZNF397 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC13250, ZNF47, ZSCAN15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~31.1 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF397(84307)

General description

Zinc finger protein 397 (ZNF397) belongs to the Cys2His2 group of the zinc-finger superfamily. It possesses a leucine-rich SCAN domain and nine Cys2His2 zinc fingers. The gene encoding this protein is localized on human chromosome 18q12.

Immunogen

ZNF397 (NP_115723.1, 1 a.a. ~ 275 a.a) full-length human protein.

Sequence
MAVESGVISTLIPQDPPEQELILVKVEDNFSWDEKFKQNGSTQSCQELFRQQFRKFCYQETPGPREALSRLQELCYQWLMPELHTKEQILELLVLEQFLSILPEELQIWVQQHNPESGEEAVTLLEDLEREFDDPGQQVPASPQGPAVPWKDLTCLRASQESTDIHLQPLKTQLKSWKPCLSPKSDCENSETATKEGISEEKSQGLPQEPSFRGIKLSRPPKASSAIRWECVSPGSFPGDIIAAEATHSTISCFAINTLPATILPSKNVNRKYFS

Biochem/physiol Actions

Zinc finger protein 397 (ZNF397) has been shown to co-localize with centromere protein-A (CENP-A) during early prophase and interphase. It might be involved in transcriptional regulation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Bailey et al.
Chromosoma, 117(4), 367-380 (2008-03-29)
The centromere is a complex structure required for equal segregation of newly synthesised sister chromatids at mitosis. One of the significant objectives in centromere research is to determine the complete repertoire of protein components that constitute the kinetochore. Here, we
Yimin Wu et al.
Gene, 310, 193-201 (2003-06-13)
We have identified two novel human SCAN domain genes ZNF396 and ZNF397, which are clustered within the region of chromosome 18q12. Three isoforms of ZNF396 transcript, 1.5, 2.5 and 3.9-kb, are expressed highly in liver. Four isoforms of ZNF397 transcript

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service