Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407170

Sigma-Aldrich

Anti-SAMD4A antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp434H0350, KIAA1053, SAMD4, SMG, SMGA, Smaug, Smaug1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~27.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SAMD4A(23034)

General description

Sterile alpha motifs (SAMs) in proteins such as SAMD4A are part of an RNA-binding domain that functions as a posttranscriptional regulator by binding to an RNA sequence motif known as the Smaug recognition element, which was named after the Drosophila Smaug protein (Baez and Boccaccio, 2005 [PubMed 16221671]).[supplied by OMIM

Immunogen

SAMD4A (AAH57838.1, 1 a.a. ~ 247 a.a) full-length human protein.

Sequence
MMFRDQVGVLAGWFKGWNECEQTVALLSLLKRVSQTQARFLQLCLEHSLADCAELHVLEREANSPGIINQWQQESKDKVISLLLTHLPLLKPGNLDAKVEYMKLLPKTLAHSIEHNQHIEESRQLLSYALIHPATSLEDRSALAMWLNHLEDRTSTSFGGQNRGRSDSVDYGQTHYYHQRQNSDDKLNGWQNSRDSGICINASNWQDKSMGCENGHVPLYSSSSVPTTINTIGTSTSTSKFPGIPLT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soon Woo Nam et al.
International journal of oncology, 45(4), 1450-1456 (2014-07-16)
Hepatocellular carcinoma (HCC) is an aggressive malignancy with a poor prognosis and a very complex dysregulated molecular etiology. Furthermore, conventional therapy thus far has had only limited success. A recently developed oral multikinase inhibitor, sorafenib, has been used to improve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service