Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1406363

Sigma-Aldrich

Anti-RAD17 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CCYC, FLJ41520, HRAD17, R24L, RAD17SP, RAD24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD17(5884)

General description

The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad17, a cell cycle checkpoint gene required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein shares strong similarity with DNA replication factor C (RFC), and can form a complex with RFCs. This protein binds to chromatin prior to DNA damage and is phosphorylated by the checkpoint kinase ATR following damage. This protein recruits the RAD1-RAD9-HUS1 checkpoint protein complex onto chromatin after DNA damage, which may be required for its phosphorylation. The phosphorylation of this protein is required for the DNA-damage-induced cell cycle G2 arrest, and is thought to be a critical early event during checkpoint signaling in DNA-damaged cells. Eight alternatively spliced transcript variants of this gene, which encode four distinct proteins, have been reported. Two pseudogenes, located on chromosomes 7 and 13, have been identified. (provided by RefSeq)

Immunogen

RAD17 (NP_579919.1, 1 a.a. ~ 584 a.a) full-length human protein.

Sequence
MNQHELAVHKKKIEEVETWLKAQVLERQPKQGGSILLITGPPGCGKTTTLKILSKEHGIQVQEWINPVLPDFQKDDFKGMFNTESSFHMFPYQSQIAVFKEFLLRATKYNKLQMLGDDLRTDKKIILVEDLPNQFYRDSHTLHEVLRKYVRIGRCPLIFIISDSLSGDNNQRLLFPKEIQEECSISNISFNPVAPTIMMKFLNRIVTIEANKNGGKITVPDKTSLELLCQGCSGDIRSAINSLQFSSSKGENNLRPRKKGMSLKSDAVLSKSKRRKKPDRVFENQEVQAIGGKDVSLFLFRALGKILYCKRASLTELDSPRLPSHLSEYERDTLLVEPEEVVEMSHMPGDLFNLYLHQNYIDFFMEIDDIVRASEFLSFADILSGDWNTRSLLREYSTSIATRGVMHSNKARGYAHCQGGGSSFRPLHKPQWFLINKKYRENCLAAKALFPDFCLPALCLQTQLLPYLALLTIPMRNQAQISFIQDIGRLPLKRHFGRLKMEALTDREHGMIDPDSGDEAQLNGGHSAEESLGEPTQATVPETWSLPLSQNSASELPASQPQPFSAQGDMEENIIIEDYESDGT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service