Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405458

Sigma-Aldrich

Anti-ANXA3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ANX3

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~36.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

General description

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation. (provided by RefSeq)

Immunogen

ANXA3 (NP_005130.1, 1 a.a. ~ 323 a.a) full-length human protein.

Sequence
MASIWVGHRGTVRDYPDFSPSVDAEAIQKAIRGIGTDEKMLISILTERSNAQRQLIVKEYQAAYGKELKDDLKGDLSGHFEHLMVALVTPPAVFDAKQLKKSMKGAGTNEDALIEILTTRTSRQMKDISQAYYTVYKKSLGDDISSETSGDFRKALLTLADGRRDESLKVDEHLAKQDAQILYKAGENRWGTDEDKFTEILCLRSFPQLKLTFDEYRNISQKDIVDSIKGELSGHFEDLLLAIVNCVRNTPAFLAERLHRALKGIGTDEFTLNRIMVSRSEIDLLDIRTEFKKHYGYSLYSAIKSDTSGDYEITLLKICGGDD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Yong Wu et al.
Medical oncology (Northwood, London, England), 31(8), 108-108 (2014-07-10)
Autophagy is a cellular recycling process to enable cell survival in less favorable conditions through degradation of their unnecessary cellular components and utilization of the breakdown components. Autophagy also plays an important role in tumor pathology. In this study, we
Cheng Yang et al.
Biochimica et biophysica acta, 1842(11), 2306-2317 (2014-09-16)
Helix B surface peptide (HBSP), derived from erythropoietin, displays powerful tissue protection during kidney ischemia reperfusion (IR) injury without erythropoietic side effects. We employed cyclization strategy for the first time, and synthesized thioether-cyclized helix B peptide (CHBP) to improve metabolic
Conghong Shi et al.
Biochemical and biophysical research communications, 452(3), 308-314 (2014-08-15)
Recent research has revealed a role for Ambra1, an autophagy-related gene-related (ATG) protein, in the autophagic pro-survival response, and Ambra1 has been shown to regulate Beclin1 and Beclin1-dependent autophagy in embryonic stem cells and cancer cells. However, whether Ambra1 plays
Teng Jiang et al.
British journal of pharmacology, 171(13), 3146-3157 (2014-03-13)
Recent clinical trials report that metformin, an activator of AMP-activated protein kinase (AMPK) used to treat type 2 diabetes, significantly reduces the risk of stroke by actions that are independent of its glucose-lowering effects. However, the underlying molecular mechanisms are
Urmi Bandyopadhyay et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(30), 11055-11060 (2014-07-16)
Lipofuscin, or aging pigment, is accreted as red autofluorescence in the lysosomes of motor neuron cell bodies in the ventral horn of WT mice by 3 mo of age. Strikingly, in two presymptomatic ALS mouse strains transgenic for mutant human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service