SAB1405213
Monoclonal Anti-MTUS1 antibody produced in mouse
clone 1C10, ascites fluid
Synonym(s):
ATIP, DKFZp586D1519, DKFZp686F20243, FLJ14295, KIAA1288, MP44, MTSG1
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
1C10, monoclonal
mol wt
antigen ~52.51 kDa
species reactivity
human
technique(s)
indirect ELISA: suitable
western blot: 1:500-1:1000
isotype
IgMκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MTUS1(57509)
General description
Immunogen
Sequence
MQLQEQFDNLNAAHETSKLEIEASHSEKLELLTKAYEASLSEIKKGHEIEKKSLEDLLSEKQESLEKQINDLKSENDALNEKLKSEEQKRRAREKANLKNPQIMYLEQELESLKAVLEIKNEKLHQQDIKLMKMGKLVDNNTALVDKLKRFQQENEELKARMDKHMAISRQLSTEQAVLQESLEKESKVNKRLSMENEELLWKLHNGDLCSPKRSPTSSAIPLQSPRNSGSFPSPSISPR
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service