Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405101

Sigma-Aldrich

Monoclonal Anti-PTPLAD1, (N-terminal) antibody produced in mouse

clone 5B5, ascites fluid

Synonym(s):

B-IND1, FLJ90376, HSPC121

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5B5, monoclonal

mol wt

antigen ~38.54 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG1κ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPLAD1(51495)

Immunogen

PTPLAD1 (NP_057479, 1 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MENQVLTPHVYWAQRHRELYLRVELSDVQNPAISITENVLHFKAQGHGAKGDNVYEFHLEFLDLVKPEPVYKLTQRQVNITVQKKVSQWWERLTKQEKRPLFLAPDFDRWLDE

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service