SAB1404056
Monoclonal Anti-MCM5 antibody produced in mouse
clone 2H8, ascites fluid
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
2H8, monoclonal
mol wt
antigen ~106.85 kDa
species reactivity
human
technique(s)
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MCM5(4174)
General description
Immunogen
Sequence
MSGFDDPGIFYSDSFGGDAQADEGQARKSQLQRRFKEFLRQYRVGTDRTGFTFKYRDELKRHYNLGEYWIEVEMEDLASFDEDLADYLYKQPAEHLQLLEEAAKEVADEVTRPRPSGEEVLQDIQVMLKSDASPSSIRSLKSDMMSHLVKIPGIIIAASAVRAKATRISIQCRSCRNTLTNIAMRPGLEGYALPRKCNTDQAGRPKCPLDPYFIMPDKCKCVDFQTLKLQELPDAVPHGEMPRHMQLYCDRYLCDKVVPGNRVTIMGIYSIKKFGLTTSRGRDRVGVGIRSSYIRVLGIQVDTDGSGRSFAGAVSPQEEEEFRRLAALPNVYEVISKSIAPSIFGGTDMKKAIACLLFGGSRKRLPDGLTRRGDINLLMLGDPGTAKSQLLKFVEKCSPIGVYTSGKGSSAAGLTASVMRDPSSRNFIMEGGAMVLADGGVVCIDEFDKMREDDRVAIHEAMEQQTISIAKAGITTTLNSRCSVLAAANSVFGRWDETKGEDNIDFMPTILSRFDMIFIVKDEHNEERDVMLAKHVITLHVSALTQTQAVEGEIDLAKLKKFIAYCRVKCGPRLSAEAAEKLKNRYIIMRSGARQHERDSDRRSSIPITVRQLEAIVRIAEALSKMKLQPFATEADVEEALRLFQVSTLDAALSGTLSGVEGFTSQEDQEMLSRIEKQLKRRFAIGSQVSEHSIIKDFTKQKYPEHAIHKVLQLMLRRGEIQHRMQRKVLYRLK
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service