Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1403492

Sigma-Aldrich

Monoclonal Anti-RASSF3 antibody produced in mouse

clone 3A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC119194, MGC119195, MGC119197, RASSF5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.35 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASSF3(283349)

General description

The RAS oncogene (MIM 190020) is mutated in nearly one-third of all human cancers. Members of the RAS superfamily are plasma membrane GTP-binding proteins that modulate intracellular signal transduction pathways. A subfamily of RAS effectors, including RASSF3, share a RAS association (RA) domain.[supplied by OMIM

Immunogen

RASSF3 (NP_835463.1, 29 a.a. ~ 112 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QGKPRSGQQDVEKEKETHSYLSKEEIKEKVHKYNLAVTDKLKMTLNSNGIYTGFIKVQMELCKPPQTSPNSGKLSPSSNGCMNT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service