SAB1402991
Monoclonal Anti-BLZF1, (C-terminal) antibody produced in mouse
clone 2E10, purified immunoglobulin, buffered aqueous solution
Synonym(s):
GOLGIN-45, JEM-1, JEM-1s, JEM1, MGC22497
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2E10, monoclonal
form
buffered aqueous solution
mol wt
antigen ~37.11 kDa
species reactivity
human
technique(s)
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... BLZF1(8548)
Related Categories
General description
The gene BLZF1 (basic leucine zipper nuclear factor 1) is mapped to human chromosome 1q24. It belongs to the b-ZIP family of proteins. The encoded protein has a PEST (proline [P], glutamic acid [E], serine [S], and threonine [T]) motif, a putative leucine repeat and several phosphorylation sites. This protein is present in the nucleus, cytoplasm and Golgi apparatus.
Immunogen
Sequence
KLAKAVNSHLLGNVGINNQKKIPSTVEFCSTPAEKMAETVLRILDPVTCKESSPDNPFFESSPTTLLATKKNIGRFHPYTRYENITFNCCNHCRGELIAL
Biochem/physiol Actions
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service