Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1400236

Sigma-Aldrich

Anti-RAD23B antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HHR23B, Anti-HR23B, Anti-P58

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD23B(5887)

Related Categories

General description

The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells. (provided by RefSeq)

Immunogen

RAD23B (AAH20973, 1 a.a. ~ 409 a.a) full-length human protein.

Sequence
MQVTLKTLQQQTFKIDIDPEETVKALKEKIESEKGKDAFPVAGQKLIYAGKILNDDTALKEYKIDEKNFVVVMVTKPKAVSTPAPATTQQSAPASTTAVTSSTTTTVAQAPTPVPALAPTSTPASITPASATASSEPAPASAAKQEKPAEKPAETPVATSPTATDSTSGDSSRSNLFEDATSALVTGQSYENMVTEIMSMGYEREQVIAALRASFNNPDRAVEYLLMGIPGDRESQAVVDPPQAASTGVPQSSAVAAAAATTTATTTTTSSGGHPLEFLRNQPQFQQMRQIIQQNPSLLPALLQQIGRENPQLLQQISQHQEHFIQMLNEPVQEAGGQGGGGGGGSGGIAEAGSGHMNYIQVTPQEKEAIERLKALGFPEGLVIQAYFACEKNENLAANFLLQQNFDED

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annett Linge et al.
Journal of proteome research, 13(7), 3212-3222 (2014-06-05)
Identification of protein targets that play a role in breast cancer invasion may help to understand the rapid progression of cancer and may lead to the development of new biomarkers for the disease. In this study, we compared two highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service