Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1309056

[KO Validated]Anti-LIN28B antibody produced in mouse

new

[Knockout Validated]purified monoclonal antibody, buffered aqueous solution

Synonym(s):

Protein lin-28 homolog B, Lin-28B, LIN28B, CSDD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Human)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

form

liquid

mol wt

27084Da

species reactivity

human

technique(s)

western blot: 1:500-1:2000

isotype

IgG1, kappa

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... LIN28B(389421)

General description

Acts as a suppressor of microRNA (miRNA) biogenesis by specifically binding the precursor let-7 (pre-let-7), a miRNA precursor. Acts by binding pre-let-7 and recruiting ZCCHC11/TUT4 uridylyltransferase, leading to the terminal uridylation of pre- let-7. Uridylated pre-let-7 miRNAs fail to be processed by Dicer and undergo degradation. Specifically recognizes the 5′-GGAG-3′ motif in the terminal loop of pre-let-7. Also recognizes and binds non pre-let-7 pre-miRNAs that contain the 5′-GGAG-3′ motif in the terminal loop, leading to their terminal uridylation and subsequent degradation. Mediates MYC-mediated let-7 repression. Isoform 1, when overexpressed, stimulates growth of the breast adenocarcinoma cell line MCF-7. Isoform 2 has no effect on cell growth.

Specificity

This LIN28B antibody is generated from a mouse immunized with a recombinant protein of human LIN28B.

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein G column, followed by dialysis against PBS.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service