SAB1309023
Anti-phospho-mouse TSC1(S591) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution
Synonym(s):
Hamartin, Tuberous sclerosis 1 protein homolog, Tsc1, Kiaa0243
About This Item
biological source
(Mouse)
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
mol wt
128746Da
species reactivity
mouse
technique(s)
dot blot: 1:500
isotype
IgG
NCBI accession no.
storage temp.
-10 to -25°C
target post-translational modification
phosphorylation (pSer591)
Gene Information
mouse ... Tsc1(64930)
General description
Specificity
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
SKU | GTIN |
---|---|
SAB1309023-400UL | 4065272322515 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service