Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1309010

Anti-phospho-mouse KIT(T357) antibody produced in rabbit

new

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Mast/stem cell growth factor receptor Kit, SCFR, Proto-oncogene c-Kit, Tyrosine-protein kinase Kit, CD117, Kit, Sl

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

(Mouse)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

109343Da

species reactivity

mouse

technique(s)

dot blot: 1:500

isotype

IgG

NCBI accession no.

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pThr357)

Gene Information

mouse ... Kit(16590)

General description

The c-Kit proto-oncogene is the cellular homolog of thetransforming gene of a feline retrovirus (v-Kit). The c-kit proteinincludes characteristics of a protein kinase transmembranereceptor. Alternatively spliced transcript variants encodingdifferent isoforms have been found for this gene. [provided byRefSeq].

Specificity

This mouse KIT Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T357 of mouse KIT.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service