Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300429

Sigma-Aldrich

Anti-MINK1/2 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GCK family kinase MiNK, Anti-MAP4K6, Anti-MAPK/ERK kinase kinase kinase 6, Anti-Misshapen/NIK-related kinase, Anti-Mitogen-activated protein kinase kinase kinase kinase 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MINK1(50488)

General description

MINK1 and MINK2 are serine/threonine protein kinases of the STE subfamily that may play a role in the response to environmental stress. They appears to act upstream of the JUN N-terminal pathway and may play a role in the development of the brain. Expression appears to be ubiquitous, highly expressed in the brain, moderately expressed in kidney and spleen, with lower levels present in heart and skeletal muscle. These proteins are up-regulated during post-natal brain development.

Immunogen

MINK1/2 (Q8N4C8 (-2 or -3), 681-716)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human MINK1/2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service