Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRS3771

Sigma-Aldrich

Anti-HRK antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Harakiri, Anti-Harakiri, BCL2 Interacting protein (contains only BH3 domain), Anti-Neuronal death proteinDP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 10 kDa

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HRK(8739)

Related Categories

Immunogen

a 15 amino acid peptide from near the center of human Hrk.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3771.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinnosuke Yahiro et al.
Cell death discovery, 7(1), 360-360 (2021-11-21)
Locus for Enterocyte Effacement (LEE)-positive Shiga-toxigenic Escherichia coli (STEC) contributes to many global foodborne diseases, with infection characterized by severe gastrointestinal symptoms, including bloody diarrhea. The incidence of LEE-negative STEC-mediated disease is also increasing globally. Subtilase cytotoxin (SubAB) is released
Jessica F Boehler et al.
The Journal of pathology, 249(2), 215-226 (2019-05-29)
The etiology of myositis is unknown. Although attempts to identify viruses in myositis skeletal muscle have failed, several studies have identified the presence of a viral signature in myositis patients. Here we postulate that in individuals with susceptible genetic backgrounds
Erica Anspach Will et al.
Biology of reproduction, 96(4), 843-854 (2017-04-04)
The present studies were designed to determine whether progesterone (P4)-progesterone receptor membrane component 1 (PGRMC1) signaling is able to attenuate the apoptotic effects of oxidative stress induced by hydrogen peroxide (H2O2). To achieve this goal, freshly isolated human granulosa/luteal cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service