Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7373

Sigma-Aldrich

PAK7 Active human

≥75% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Synonym(s):

PAK5, p21 activated kinase-7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

assay

≥75% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

10 units/mg protein (Bradford)

mol wt

protein apparent mol wt ~116 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PAK7(57144)

Biochem/physiol Actions

PAK7 is a PAK family kinase that contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK7 has been implicated in the regulation of cell morphology, motility and transformation. PAK7 is moderately expressed in adult and fetal whole brain and in all specific adult brain regions tested.

Unit Definition

One unit will incorporate one nanomole of phosphate into the peptide substrate CKRPRAASFAE per minute at 30 °C and pH 7.2 using a final concentration of 50 μM [32P] ATP.

Physical form

Solution in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, and 30% glycerol.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service