P2122
Anti-phospho-Dynamin (pSer778) antibody produced in sheep
affinity isolated antibody, aqueous glycerol suspension
About This Item
biological source
sheep
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
aqueous glycerol suspension
species reactivity
rat, mouse, human
technique(s)
western blot: 1:1000 using rat brain synaptosomal lysate.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer778)
Gene Information
human ... DNM1(1759) , DNM2(1785) , DNM3(26052)
mouse ... Dnm1(13429) , Dnm2(13430) , Dnm3(103967)
rat ... Dnm1(140694) , Dnm2(25751) , Dnm3(171574)
Immunogen
Physical form
Not finding the right product?
Try our Product Selector Tool.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service