Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0033

Sigma-Aldrich

ProTx-II

≥95% (HPLC)

Synonym(s):

PT-II, ProToxin-II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C168H250N46O41S8
Molecular Weight:
3826.59
UNSPSC Code:
51111800

assay

≥95% (HPLC)

form

lyophilized solid

storage condition

under inert gas

color

white

storage temp.

−20°C

Biochem/physiol Actions

ProTx-II is a potent, selective Nav1.7 blocker.
ProTx-II is a potent, selective Nav1.7 blocker. Voltage-gated sodium channels (Nav) are responsible for transducing extracellular signals into action potentials in excitable cells. The Nav1 family consists of nine sub-types for which selective tools are highly valued. Discovery of subtype-selective tools has been challenging due to conservation of putative binding sites. Nav1.7 is expressed in sensory neurons and is critical for pain processing. ProTx-II, isolated from the tarantula Thrixopelma pruriens, is a potent blocker of Nav1 channels. Although ProTx-II blocks several members of this channel family, it has >100-fold selectivity for Nav1.7. ProTx-II is active in an in vitro model of pain processing. ProTx-II binds to a previously unknown site on the channel, conferring selectivity and increasing its value as a research tool. It blocks Nav1.7 by decreasing conductance and shifting the activation potential in the positive direction, resulting in an overall decrease in excitability.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service