Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA026751

Sigma-Aldrich

Anti-TRMT61B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Potential tRNA, Anti-adenine-N(1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GVCAVYVVNITQVIELLDGIRTCELALSCEKISEVIVRDWLVCLAKQKNGILAQKVESKINTDVQLDSQEKIGVKGELFQEDDHEESHSDFPYGSFPYVARPVHWQPGHTAFLVKLRKV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRMT61B(55006)

General description

The tRNA methyltransferase 61B (TRMT61B) gene is mapped to human chromosome 2p23.2. TRMT61B protein encoded by this gene is primarily localized to mitochondria. The encoded protein forms a homotetrameric structure similar to bacterial methyltransferase (TrmI). Human Trmt61B is characterized with C-terminal Flag tag containing 334 amino acids.

Immunogen

Potential tRNA recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Methyltransferase encoded by the gene tRNA methyltransferase 61B (TRMT61B) plays a vital role in 1-methyladenosine (m1A) A58 formation in human mitochondrial tRNAs. TRMT61B catalyzes the methylation of 16S rRNA present in mitochondria and it is also involved in tRNA metabolic process. This protein is considered as the first vertebrate methyltransferase that alters both tRNA and rRNAs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73855

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial 16S rRNA Is Methylated by tRNA Methyltransferase TRMT61B in All Vertebrates.
Bar-Yaacov D
PLoS Biology, 14(9) (2016)
Sonia Fabris et al.
American journal of hematology, 88(1), 24-31 (2012-10-10)
Recent studies have described chromosome 2p gain as a recurrent lesion in chronic lymphocytic leukemia (CLL). We investigated the 2p gain and its relationship with common prognostic biomarkers in a prospective series of 69 clinical monoclonal B-cell lymphocytosis (cMBL) and
Takeshi Chujo et al.
RNA (New York, N.Y.), 18(12), 2269-2276 (2012-10-26)
In human mitochondria, 1-methyladenosine (m¹A) occurs at position 58 of tRNA(Leu(UUR)). In addition, partial m¹A58 modifications have been found in human mitochondrial tRNA(Lys) and tRNA(Ser(UCN)). We identified human Trmt61B, which encodes a mitochondria-specific tRNA methyltransferase responsible for m¹A58 in these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service